Heat and cut resistant gloves
Supplier: JUTEC
H0150013 H0150035
112-1962EA 65.6 EUR
112-1962 JUTEH0150035
Heat and cut resistant gloves
Gloves
These five-finger gloves in Kevlar® coarse knit with cotton insulation protect against contact heats of up to 250 °C.
  • Perfect for work involving slight heat influence
  • The gloves are single-insulated and offer excellent handling
  • Washable
  • With knitted cuff
The gloves can be used in any environment in which direct exposure to flames is ruled out from the outset.
Certifications: EN 388 (244X), EN 407 (X1XXXX); Cat. II.