Shelving systems for drums, WSC-G-E
Supplier: LACONT UMWELTTECHNIK
C22-5506-B C22-5501-B C22-5502-B C22-5503-B C22-5504-B C22-5505-B
LACOC22-5506-BEA 0 EUR
LACOC22-5506-B LACOC22-5501-B LACOC22-5502-B LACOC22-5503-B LACOC22-5504-B LACOC22-5505-B
Shelving systems for drums, WSC-G-E
Furniture Shelves
Water protection compartment containers are suitable for the storage of hazardous substances indoors.
Storage can take place both directly on the grid supports of the different levels and also on euro- or chemical pallets. The compact modular design allows an uncomplicated change of location in a very short time.