Pallets and sumps for KTC with filling stand, WSP
Supplier: LACONT UMWELTTECHNIK
P21-2603-B P21-2606-A P21-1603-B P21-1606-A P21-2608-A P21-1608-A
LACOP21-2603-BEA 0 EUR
LACOP21-2603-B LACOP21-2606-A LACOP21-1603-B LACOP21-1606-A LACOP21-2608-A LACOP21-1608-A
Pallets and sumps for KTC with filling stand, WSP
Drum Storage
These pallets and sumps with filling stand are used for storing acids and alkalis.
  • Sump capacity of 1000 L
  • With chemical resistant PE-inserts for storing acids and alkalis