Drum sumps, PE-100-EP
Supplier: LACONT UMWELTTECHNIK
P70-7006-A P70-7007-A P70-7008-A
LACOP70-7006-AEA 0 EUR
LACOP70-7006-A LACOP70-7007-A LACOP70-7008-A
Drum sumps, PE-100-EP
Drum Storage
Rotationally-molded body of black PE.
  • Circumferential stabilising ribs
  • Easily removable PE grid support
Ideal for storing water-endangering and aggressive media.