Sumps for drums and small containers, PE-BS
Supplier: LACONT UMWELTTECHNIK
P70-7070-A P70-7073-A P70-7074-A P70-7072-A P70-7071-A
LACOP70-7070-AEA 0 EUR
LACOP70-7070-A LACOP70-7073-A LACOP70-7074-A LACOP70-7072-A LACOP70-7071-A
Sumps for drums and small containers, PE-BS
Drum Storage
These sumps of rotationally-molded body of black PE are used for storing water-endangering and aggressive media and suited for covering large-scale surfaces.
  • Circumferential stabilising ribs
  • Designed for placing flat on the floor
  • Individual segments can be combined by galvanised element connectors
  • Galvanised or PE grid support
  • Maximum capacity 1000 kg/m2 with uniformly distributed load