Pressure vessels
Supplier: Merck
XX6700P20 XX6700P05 XX6700P10 XX6700P01
MILFXX6700P20EA 2370 EUR
MILFXX6700P20 MILFXX6700P05 MILFXX6700P10 MILFXX6700P01
Pressure vessels
Reaction Vessels Reaction Vessels, Cylindrical Shape
Dispensing pressure vessels hold liquid for filtration through pressure-operated filter holders, if connected to an external pressure source. Used for large-volume filtration, reservoir for buffer or solvent dispensing.
  • Made from stainless steel 316
  • Closures are secured by cam-lock handle
  • Vessels are autoclavable when vented
  • Fluoroelastomer gaskets and O-rings used
  • Molded neoprene rubber base
Max. operating pressure: 6,9 bar
Max. operating temperature: 121 °C
Certifications: Meets ASME®-UM Code requirements.