Heat resistant gloves, PBI®
Supplier: JUTEC
H111B140-PBI H111B130-PBI
JUTEH111B140-PBIEA 229 EUR
JUTEH111B140-PBI JUTEH111B130-PBI
Heat resistant gloves, PBI®
Gloves
These mittens made of PBI® fabric provide protection against contact heat up to 800 °C.
  • Cut and abrasion resistant
  • Special insulation
  • Universal size fits all
Ideal for use in foundries and in the metal industry.
Certifications: EN 388, EN 407; Cat. III