Safety ankle boots, lace-up, Bacou Original Cerata
Supplier: HONEYWELL SAFETY
6246102/43 6246102/39 6246102/40 6246102/42 6246102/44 6246102/45 6246102/46 6246102/47 6246102/48 6246102/49 6246102/41
DALL6246102/43EA 120 EUR
DALL6246102/43 DALL6246102/39 DALL6246102/40 DALL6246102/42 DALL6246102/44 DALL6246102/45 DALL6246102/46 DALL6246102/47 DALL6246102/48 DALL6246102/49 DALL6246102/41
Safety ankle boots, lace-up, Bacou Original Cerata
Shoes and Boots
Safety shoe made from water resistant, oiled, full grain leather upper with textile panels, 3-D Lining plus injected, dual-density PU outsole.
  • Reinforced protection at the front and back
  • Padded ankle collar and tongue
  • Antibacterial, breathable, quick drying lining with 3-D structure
  • Removable, antibacterial, dual-density PU foam insole with comfortable heel pads
  • Lateral watertight bellows prevent liquid and dust from entering boots
  • Light, metal-free Spacium™ toecap in injected polymer (200 J)
  • Stainless steel puncture resistant midsole
For diverse environments: Frozen ground and very high temperatures.
Colour: Black/green
Certifications: EN ISO 20345:2011; S3, SRC, CI, HO