Demecolcine solution_SC 10 µg/ml PBS
Supplier: BIOWEST
Synonyms: Colcemid
L0040-050 L0040-020 L0040-010
L0040-050EA 34 EUR
L0040-050 L0040-020 L0040-010
Demecolcine solution_SC 10 µg/ml PBS
Demecolcine
Colcemid arrests the division of cells in mitosis. It prevents the formation of the spindle apparatus responsible for cell division, thereby permitting an accumulation of metaphases.
Formula: C₂₁H₂₅NO₅
MW: 371.433 g/mol
MDL Number: MFCD00075459
CAS Number: 477-30-5