Bottles, without pump vaporiser, BOSTON
Supplier: Müller + Krempel
MUEL14.289.20EA 16.2 EUR
MUEL14.289.20
Bottles, without pump vaporiser, BOSTON
Bottles Spray Bottles
Cosmetic bottles, made of PET, clear.
  • Good chemical resistance
  • Good temperature resistance
  • Pump vaporiser to be ordered separately
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904