Analytical HPLC columns, XTerra® MS C8
Supplier: WATERS
186000433 186000399
WATE186000433EA 1590 EUR
WATE186000433 WATE186000399
Analytical HPLC columns, XTerra® MS C8
Chromatography Columns
XTerra® MS C8 columns are general purpose, hybrid-based, reversed phase C8 columns that are designed to be compatible with mass spectrometry applications. These first generation columns provide superior pH stability compared to a silica based column.