ESD safety shoes, lace-up, GX 120
Supplier: ATLAS - the shoe company
53035 53036 53037 53038 53039 53040 53041 53042 53043
114-4651EA 0 EUR
114-4651 ATLA53036 ATLA53037 ATLA53038 ATLA53039 ATLA53040 ATLA53041 ATLA53042 ATLA53043
ESD safety shoes, lace-up, GX 120
Shoes and Boots
ESD safety shoes made from waterproof leather and a MPU® Light sole (3,5 mm profile, oil, acid and petrol resistant, anti-static).
  • Skin friendly, abrasion resistant aktiv-X lining for enhanced breathability
  • Removable insoles; orthopedic insoles possible
  • 3D cushioning system: pressure relieving insole, cushioning comfort midsole, dynamic outsole system made from MPU
  • Protective steel toecap (200 J)
  • Electrical contact resistance of 1×10⁵ to 3,5×10⁷ Ohm
Colour: Black
Certifications: EN 61340 (ESD); DGUV 112-191; EN ISO 20345; S2