ESD safety shoes, sandals, GX 350
Supplier: ATLAS - the shoe company
45500/36 45500/39 45500/40 45500/41 45500/42 45500/43 45500/38 45500/37
114-4662EA 206 EUR
114-4662 114-4665 114-4666 114-4667 114-4668 114-4669 114-4664 114-4663
ESD safety shoes, sandals, GX 350
Shoes and Boots
ESD safety sandals made from Sportline upper (perforated oiled nubuck leather) and a MPU® Light sole (3,5 mm profile, oil, acid and petrol resistant, anti-static).
  • Moisture control and breathability thanks to Clima-stream® technology
  • Removable insoles; orthopedic insoles possible
  • 3D cushioning system: pressure relieving insole, cushioning comfort midsole, dynamic outsole system made from MPU
  • Protective steel toecap (200 J)
  • Electrical contact resistance of 1×10⁵ to 3,5×10⁷ Ohm
Colour: Black
Certifications: EN 61340 (ESD); DGUV 112-191; EN ISO 20345; S1