VWR® Q Path®, Containers with Cap for Biopsy Probes
Supplier: VWR Collection
216-1310EA 46.4 EUR
216-1310 216-1319 216-1317 216-1309 216-1307 216-1315 216-1308 216-1306
VWR® Q Path®, Containers with Cap for Biopsy Probes
Sample Containers General Purpose Containers
Q Path® containers (PP) with suitable caps (PE) for safe transport of biopsy probes.
  • Different colours
  • High impermeability
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904