VWR® Q Path® Adhesion Slides
Supplier: LABONORD
10048001 10048002 10048003 10048004 10048005
630-2087EA 33.5 EUR
630-2087 630-2088 630-2089 630-2090 630-2091
VWR® Q Path® Adhesion Slides
Slides Adhesive Slides
These slides can be used for the diagnostic testing of cells suspension and tissue samples.
  • Store in a place with a stable temperature and low level humidity
Suitable for in vitro diagnostic (IVD) applications.
Certifications: EN 375:2001. ISO 8037/1.