ESD occupational shoes, sandals, Velcro® fastening, X-Light, 7131131/7131136
Supplier: Abeba
7131131/35 7131131/36 7131131/37 7131131/38 7131131/39 7131131/40 7131131/41 7131131/42 7131131/43 7131131/44 7131131/45 7131131/46 7131131/47 7131131/48 7131136/35 7131136/36 7131136/37 7131136/38 7131136/39 7131136/40 7131136/41 7131136/42 7131136/43 7131136/44 7131136/45 7131136/46 7131136/47 7131136/48
ABEB7131131/35EA 96.1 EUR
ABEB7131131/35 ABEB7131131/36 ABEB7131131/37 ABEB7131131/38 ABEB7131131/39 ABEB7131131/40 ABEB7131131/41 ABEB7131131/42 ABEB7131131/43 ABEB7131131/44 ABEB7131131/45 ABEB7131131/46 ABEB7131131/47 ABEB7131131/48 ABEB7131136/35 ABEB7131136/36 ABEB7131136/37 ABEB7131136/38 ABEB7131136/39 ABEB7131136/40 ABEB7131136/41 ABEB7131136/42 ABEB7131136/43 ABEB7131136/44 ABEB7131136/45 ABEB7131136/46 ABEB7131136/47 ABEB7131136/48
ESD occupational shoes, sandals, Velcro® fastening, X-Light, 7131131/7131136
Shoes and Boots
ESD shoe made from smooth leather with Air Mesh textile inlays and a lightweight, fuel resistant PU sole.
  • Perspiration-absorbent special lining with silver ions
  • Replaceable active comfort insole provides optimised comfort; orthopaedic insoles possible
  • Sanitized® treatment reduces odour and bacterial growth
  • Lightweight PU sole with high slip resistance and shock-absorbent heel
  • Double Velcro® fastening
Colour: White or black
Colour: White or black
Certifications: EN 61340 (ESD); DGUV 112-191; EN ISO 20347:2012; O1, FO, SRC.