Cut resistant gloves, Vis-Tex5, A625
Supplier: Portwest
A625O8RL A625O8RM A625O8RXL A625Y8RL A625Y8RM A625Y8RXL
PRTWA625O8RLEA 80 EUR
PRTWA625O8RL PRTWA625O8RM PRTWA625O8RXL PRTWA625Y8RL PRTWA625Y8RM PRTWA625Y8RXL
Cut resistant gloves, Vis-Tex5, A625
Gloves
These flexible gloves made of HPPE and glass fibre with PU coating on the palm provide cut resistance and high abrasion resistance.
  • Flexible high visibility lining for optimum protection and comfort
  • PU dipping for high abrasion resistance
Certifications: EN 388:2016 (4443DX), EN 407 (X1XXXX)