Flame retardant trousers, BizFlame™ Plus, FR56
Supplier: Portwest
FR56NARM FR56NARS FR56NARL FR56NARXL FR56NARXXL FR56NARXXXL
PRTWFR56NARMEA 87.8 EUR
PRTWFR56NARM PRTWFR56NARS PRTWFR56NARL PRTWFR56NARXL PRTWFR56NARXXL PRTWFR56NARXXXL
Flame retardant trousers, BizFlame™ Plus, FR56
Workwear
These trousers are constructed from 99% cotton interwoven with 1% carbon fibres. The fabric has flame-resistant and anti-static properties and is therefore ideal for offshore and welding industries.
  • Double stitched, industrial wash, reflective tape on lower legs for enhanced visibility
  • Concealed front brass zip
  • Back pocket, kneepad pockets
  • Adjustable hem
Weight: 350 g/m²
Certifications: EN ISO 11612 (A1+A2, B1, C1, F1), EN ISO 11611 (A1+A2, Class 2), EN 1149-5
Order Now
SPECIFICATIONS
Model FR56
Garment application Welding
Environmentally Preferable
Risk area Heat,Flame-retardant,Anti-static
Gender Unisex
Sleeve or leg length Long
Closure type Zip closure
Fabric 99% cotton interwoven with 1% carbon fibres
Weight 350 g/m²