Chest wader, Steelite™ Safety S5, FW74
Supplier: Portwest
FW74BKR43 FW74BKR46 FW74BKR44 FW74BKR41 FW74BKR42 FW74BKR47 FW74BKR37 FW74BKR38 FW74BKR39 FW74BKR40 FW74BKR45
PRTWFW74BKR43EA 71 EUR
PRTWFW74BKR43 PRTWFW74BKR46 PRTWFW74BKR44 PRTWFW74BKR41 PRTWFW74BKR42 PRTWFW74BKR47 PRTWFW74BKR37 PRTWFW74BKR38 PRTWFW74BKR39 PRTWFW74BKR40 PRTWFW74BKR45
Chest wader, Steelite™ Safety S5, FW74
Workwear
Heavy-duty PVC/nitrile safety wader with adjustable straps, cord adjustable waist, stainless steel eyelets and integrated S5 safety boots.
  • Colour-coded red outsole for easy identification of safety features
  • Slip-and oil resistant, anti-static outsole
  • Energy absorber heel
  • Toe protection cap made from steel (200 J)
  • Pierce resistant steel midsole
Colour: Black
EN ISO 20345:2011, S5
Certifications: EN ISO 20345:2011, S5