Activated charcoal filter papers, grade MN 728
Supplier: MACHEREY-NAGEL
481009 481011 481012 481018 481005 481007 481015 481024 481032 4810101 4810047
516-6018EA 24.9 EUR
516-6018 MANA481011 MANA481012 MANA481018 MANA481005 MANA481007 MANA481015 MANA481024 MANA481032 MANA4810101 MANA4810047
Activated charcoal filter papers, grade MN 728
Filters Filter Papers Qualitative Filter Paper
The activated charcoal filter paper MN 728 is particularly suited for the clarification and decolouring of solutions. The activated charcoal is incorporated in the paper and cannot be washed out into the filtrate.