Jars, with slip lid
Supplier: SEMADENI
1992 1991 3998 1993 1997 2962 3395
216-3160EA 14 EUR
216-3160 SEMA1991 SEMA3998 SEMA1993 SEMA1997 SEMA2962 SEMA3395
Jars, with slip lid
Jars General Use Jars
PS, clear
  • Good chemical resistance
  • Temperature resistance from −10 to 70 °C
Ideal for sampling and storage of solid samples.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904