Capillary columns, DC-200
Supplier: Agilent
G3591-81027 G3591-81032 G3591-81039 G3591-81082 G3591-81140 G3591-82001 G3591-82032 G3591-82039 G3591-82140 G3591-82082 G3591-80001 G3591-80027 G3591-80030 G3591-80032 G3591-80039 G3591-80082 G3591-80140 G3591-81001
HEWLG3591-81027EA 0 EUR
HEWLG3591-81027 HEWLG3591-81032 HEWLG3591-81039 HEWLG3591-81082 HEWLG3591-81140 HEWLG3591-82001 HEWLG3591-82032 HEWLG3591-82039 HEWLG3591-82140 HEWLG3591-82082 HEWLG3591-80001 HEWLG3591-80027 HEWLG3591-80030 HEWLG3591-80032 HEWLG3591-80039 HEWLG3591-80082 HEWLG3591-80140 HEWLG3591-81001
Capillary columns, DC-200
Chromatography Columns
Packed GC columns are designed and manufactured to offer excellent and reproducible performance for all sample types associated with packed column separations, most important in the hydrocarbon processing industry
  • Highly efficient and rigorous packing technology assures column-to-column reproducibility and ultimate efficiency
  • UltiMetal treated stainless steel tubing allows for improved inertness and peak shape performance