Safety shoes, lace-up, A 100
Supplier: ATLAS - the shoe company
A100/42 A100/43
ATLAA100/42EA 0 EUR
ATLAA100/42 ATLAA100/43
Safety shoes, lace-up, A 100
Shoes and Boots
Sneaker style safety shoes made from full-grained calf leather and a Sneaker Line sole (3,0 mm profile, oil, acid and petrol resistant, anti-static).
  • Moisture control and breathability thanks to Clima-stream® technology
  • Skin friendly, abrasion resistant aktiv-X lining for enhanced breathability
  • Removable insoles; orthopaedic insoles possible
  • 3D cushioning system: pressure relieving insole, cushioning comfort midsole, dynamic outsole system made from MPU
  • Protective steel toecap (200 J)
Colour: Brown
Certifications: DGUV 112-191; EN ISO 20345; S2