Hand disinfectants, Sterillium® Med
Supplier: Bode
981113 981112 981114 9813162_U
BODL981113EA 8.8 EUR
BODL981113 BODL981112 BODL981114 BODE9813162_U
Hand disinfectants, Sterillium® Med
Skincare Skin Disinfectants Liquid and Foam Skin Disinfectants
Sterillium® med is an ethanol based hand disinfectant designed for hygienic and surgical rub-in hand disinfection with good skin tolerability.
  • Active against bacteria, fungi and enveloped viruses
  • 1,5 min of application time for surgical hand disinfection
  • Independent of washbasin and water
  • Colourant- and fragrance-free
Caution: Use biocides safely. Always read the label and product information before use.
* These articles are only available in Germany.