Disinfectant detergent, Divodes FG VT29 L
Supplier: Diversey
7511257 100861468 G11661
DIVY7511257EA 131 EUR
DIVY7511257 141-1448 DIVYG11661
Disinfectant detergent, Divodes FG VT29 L
Cleaning Agents Disinfectants
Divodes FG a detergent disinfectant based on a mixture of propanols is specially formulated for use on surfaces that come into contact with food. It is also used on surfaces where contact with water is to be avoided.
  • Ready to use
  • Recommended for areas which are moisture-sensitive
  • Non corrosive and quick drying
  • No need to rinse after application
Certifications: Bactericide EN 1276; Fungicide EN 1650.
Caution: Use biocides safely. Always read the label and product information before use.