Disinfectant liquid, terralin® protect
Supplier: SCHUELKE MAYR
181612 181604
SCUE181612EA 112 EUR
SCUE181612 SCUE181604
Disinfectant liquid, terralin® protect
Cleaning Agents Disinfectants
Liquid concentrate for disinfection and cleaning based on a combination of aromatic alcohols, quaternary ammonium compounds, amphoteric glycine derivatives, and non ionic surfactants.
  • Broad spectrum of anti-microbial activity
  • Pleasant and fresh fragrance
  • Excellent material compatibility including sensitive materials such as acrylic glass (incubators)
Certifications: Bactericide: EN 13697; fungicidal EN 1276, EN 1650; virucidal (e. g. hepatitis B, HIV, rotaviruses and adenoviruses).
Caution: Use biocides safely. Always read the labelMore Product Information