VWR®, PCR Workstation
Supplier: VWR Collection
732-2541EA 3220 EUR
732-2541 732-2542 732-2840 732-3900
VWR®, PCR Workstation
Fume Hoods and Enclosures PCR Workstation Enclosures
The VWR® PCR Workstation is a PCR enclosure for PCR sample preparation and other sensitive protocols. It offers dual decontamination action by UV inactivation of airborne and surface-bound contaminants and, therefore, represents an ideal environment for PCR set up. Stainless steel housing, front and side panels made of 8 mm Makrolon® protects from UV irradiation.
  • Active decontamination of work surface during non working time by UV irradiation
  • Additional inactivation of aerosol-bound contaminants by shielded UV Air Recirculator during operation
  • Contaminant prevention thanks to antimicrobial stainless steel work surface
  • Function indicator for UV Air Recirculator tube
  • Displays operating time of UV tubes allowing timely replacement for constant UV intensity
  • Removable shelves provide additional storage space for reaction tubes, pipettes or racks on the rear panel
  • Four internal power outlets for operating laboratory equipment, such as mini centrifuges or vortexers, allow the combination of several working steps without interruption of the workflow
  • Electromagnetic safety mechanism stops UV irradiation if front panel is opened
Large area for stress-free working: Provides ample space and the possibility to accommodate bench top equipment, the VWR® PCR Workstation allows the combination of several working steps without change of location, thereby minimising the risk of cross-contamination.
Please refer to the manual to select the compatible Plug and Socket type for your country.
TÜV-tested, certified safety ; made in Germany.
Certifications: TÜV tested, safety certified. Made in Germany.
Delivery information: Flat Pack format for easy and convenient delivery - to be assembled.