Cleaning disinfectant, concentrate, Kohrsolin® FF
Supplier: Bode
9804320 980428
BODL9804320EA 2440 EUR
BODL9804320 BODE980428
Cleaning disinfectant, concentrate, Kohrsolin® FF
Cleaning Agents Disinfectants
Kohrsolin® FF is suitable for the prophylactic cleaning wipe disinfection of all surfaces in areas relevant to hygiene. Also suitable for targeted disinfection measures requiring an extended spectrum of activity (e.g. virucidal activity).
  • Contains glutaraldehyde 50 mg/g, benzyl-C12-18-alkyldimethyl-ammonium chlorides 30 mg/g, didecyldimethylammonium chloride 30 mg/g
  • Good cleaning properties and material compatibility
  • Bactericidal, fungicidal, tuberculocidal, sporicidal (against Clostridium difficile spores) and virucidal
  • Typical dosage: 1 ml/litre to 40 ml/litre depending on application
  • pH value of 1% solution: approx 7
Certifications: Bactericidal: EN 1276, EN 13727; Fungicidal EN 1275, EN 13624, EN 1650; Virucidal: EN 14476.
Caution: Use biocides safely. Always read the label and product information before use.