Pycnometers for bituminous materials
Supplier: NEUBERT VOLUME GLASSWAERE
0321-60-60 0321-50-1000 0321-50-0011 0321-51-1000
NEUB0321-60-60EA 169 EUR
NEUB0321-60-60 NEUB0321-50-1000 NEUB0321-50-0011 NEUB0321-51-1000
Pycnometers for bituminous materials
Specific Gravity Bottles
Soda-lime glass or borosilicate glass 3.3.
  • Two-part assembly both parts with identification number
  • Cap with funnel for ease of filling
  • Ring mark on cap stem