Tween® 80 (Polysorbate)
Supplier: G Bioscienes
786-519 786-520
786-519EA 174 EUR
786-519 786-520
Tween® 80 (Polysorbate)
Tween® 80 (Polysorbate)
Tween® 80 for solubilising membrane proteins during isolation of membrane-protein complexes.
Melting Pt: -21
MDL Number: MFCD00082107
CAS Number: 9005-65-6
Merck Index: 13,07664
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bde3ba50379784286f232046e51f1c2e5aef0684&vid=764b1ae6c7de631c0dca755bd341e627e61103f4&prid=web&clid=SAPDAM