Filter tips, LTS®
Supplier: RAININ
GP-L1000F RT-L200F RT-L1000F GP-L200F RT-L300F RT-L1200F 30389274 30389272 30389223 RT-L10FLR RT-L1000FLR 30389212 30389213 30389225 30389226 30389231 30389237 30389239 30389240 17007081 17005860 17007084 17005859 17005865
732-0260EA 269 EUR
732-0260 732-0274 613-4774 RAINGP-L200F RAINRT-L300F RAINRT-L1200F RAIN30389274 RAIN30389272 RAIN30389223 RAINRT-L10FLR RAINRT-L1000FLR RAIN30389212 RAIN30389213 RAIN30389225 RAIN30389226 RAIN30389231 RAIN30389237 RAIN30389239 RAIN30389240 RAIN17007081 RAIN17005860 RAIN17007084 RAIN17005859 RAIN17005865
Filter tips, LTS®
Pipette Tips
These aerosol-resistant tips are designed for use with RAININ Pipet-Lite, Pipet-Plus and EDP3-Plus pipettes with LTS LiteTouch Tip Ejection System. 1200 µl tips are specifically designed for use with RAININ E12-1200 and E8-1200 multi-channel pipettes. The unique design prevents drop formation and eliminates the need for touch-off between dispenses.
  • Virgin PP with PE filter, sterile
  • Filters are an extremely strong chemically inert matrix molded from pure virgin PE that does not flake or shed particles
  • Hydrophobic filter forms a barrier against aerosols and