Storage cabinets, full length
Supplier: Palbam
JG0554 JF1260C JF1260
PALBJG0554EA 5310 EUR
PALBJG0554 132-0994 132-0993
Storage cabinets, full length
Furniture Storage Cabinets
Stainless steel 304 with electropolished finish.
  • Removable shelves and sloping roof design as standard
  • Available on either castors or leveller legs
  • Assembly is quick and easy, requiring no special tools or expertise
Packaging: Typically available from stock for direct shipment from the supplier. Designed to be shipped flat-packed for self-assembly. Assembly instructions are included with all cabinets.