Test sieves, stainless steel
Supplier: LINKER INDUSTRIE TECHN
552025222016 552025222018 552025222020 552025222022 552025222024 552025222026 552025222028 552025222030 552025222032 552025222034 552025222036 552025222038 552025222040 552025222044 552025222046 552025222048 552025222050 552025222052 552025222054 552025222056 552025222058 552025222060 552025222062 552025222064 552025222066 552025222068 552025222070 552025222041 552025222059
510-9407EA 231 EUR
510-9407 510-9409 510-9411 510-9413 510-9415 510-9417 510-9419 510-9421 510-9423 510-9425 510-9427 510-9429 510-9431 510-9435 510-9437 510-9439 510-9441 510-9443 510-9445 510-9447 510-9449 510-9451 510-9453 510-9455 510-9457 510-9459 510-9461 LINK552025222041 LINK552025222059
Test sieves, stainless steel
Sieves Test Sieves
These test sieves are made from stainless steel.
  • Soft soldered seam resistant up to 170 °C
  • With identification number
Certifications: Certificate of compliance 2.1 EN 10 204. According to DIN/ISO 3310/1.