CF™ dye bovine serum albumin conjugates
Supplier: Biotium
20289 20290 20291 20292
20289.EA 331 EUR
20289. 20290. 20291. 20292.
CF™ dye bovine serum albumin conjugates
Protein Linking and Labeling Systems
Fluorescent bovine serum albumin (BSA) conjugates are commonly used as tracers in applications where physical dimensions are important. The conjugates can also be used for quantitative studies of electroporation, measurement of plasma volume in rats, and endocytosis studies. CF™ dye BSA conjugates are labelled with fluorescent CF™ dyes, which offer advantages in brightness and photostability compared to other fluorescent dyes.
Order Now
SPECIFICATIONS
Environmentally Preferable
Host
Learn more