Screwdrivers, 261 P, PicoFinish
Supplier: WIHA WERKZEUGE WILLI HAHN
00513 00512 00511 00516
238-1121EA 0 EUR
238-1121 238-1122 238-1123 238-1124
Screwdrivers, 261 P, PicoFinish
Screwdrivers
High quality chrome-vanadium-molybdenum steel blade. Ideal for fine and delicate fastening tasks for electronics, precision mechanics, model-making and hobby activities.
  • PicoFinish handle for sensitive and fast screw fixing
  • ChromTop® finish on tip for a perfect fit
  • Handle with rotating cap and rapid-turning zone