Bottles, narrow neck, with standard ground stoppers
Supplier: GLASWARENFABRIK KARL HECHT
41283036 41281012 41281006 41281018 41281024 41281030 41281036 41283006 41283012 41283018 41283024 41283030
215-1270EA 25.3 EUR
215-1270 HECH41281012 HECH41281006 HECH41281018 HECH41281024 HECH41281030 HECH41281036 HECH41283006 HECH41283012 HECH41283018 HECH41283024 HECH41283030
Bottles, narrow neck, with standard ground stoppers
Bottles Glass Bottles
Soda-lime glass, clear or amber, with standard ground joint and glass stopper
  • Good chemical and physical properties
  • For use in applications that are not associated with high thermal loads
  • Stable and robust
Amber coloured to protect light-sensitive materials.
Certifications: DIN 12036, ISO 4796