Nalgene® Carboys with Sanitary Flanges
Supplier: Thermo Fisher Scientific
2630-0020 2630-0050 2630-0010
216-1756EA 640 EUR
216-1756 216-1757 216-0304
Nalgene® Carboys with Sanitary Flanges
Bottles Carboys
PP, translucent.
  • Very good chemical resistance
  • Heat resistant up to +135 °C, autoclavable
  • Cleaning is quick and easy because there is no screw thread
With welded 3" Tri-Clover flanges for aseptic liquid transfer and/or sample collection.
Ordering information: The union clamps (734-2466) and the gaskets (734-2469), both of which are essential, must be ordered separately.