Spheroid microplates, Ultra-Low Attachment surface, Corning®
Supplier: Corning
3830 4520 4515 4516 4527
738-0203EA 2410 EUR
738-0203 738-0206 738-0204 738-0205 738-0218
Spheroid microplates, Ultra-Low Attachment surface, Corning®
Microplates Cell Culture Plates
PS, black opaque microplate with optically clear bottom, with lid. The Ultra-Low Attachment surface is a covalently bound hydrogel layer that is hydrophilic and neutrally charged. The surface minimises cell attachment, protein absorption, and enzyme activation. It is ideal for culturing stem cells, as it promotes embryoid body formation from ES cells.
  • Well geometry aids formation of spheroids in centre of well
  • Non cytotoxic, biologically inert and non degradable surface
  • Sterilised by gamma radiation