Anti-UROD Mouse Polyclonal Antibody

Supplier: Abnova
H00007389-A01
ABNOH00007389-A01EA 260 EUR
ABNOH00007389-A01
Anti-UROD Mouse Polyclonal Antibody
Antibodies
Type: Primary
Antigen: UROD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR