Anti-UVRAG Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB1136-100 ORB1136-200
BIRBORB1136-100EA 0 EUR
BIRBORB1136-100 BIRBORB1136-200
Anti-UVRAG Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:100-500; Immunochemistry: 1:50-250 (based on 0.5 mg/ml)

Type: Primary
Antigen: UVRAG (UV radiation resistance associated)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR