Anti-CCP110 Rabbit polyclonal antibody

Supplier: Biorbyt
ORB1392-100 ORB1392-200
BIRBORB1392-100EA 0 EUR
BIRBORB1392-100 BIRBORB1392-200
Anti-CCP110 Rabbit polyclonal antibody
Antibodies
Western Blot: 1:100-500; Immunochemistry: 1:50-250 (based on 0.5 mg/ml)

Type: Primary
Antigen: CCP110 (centriolar coiled coil protein 110kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR