Anti-Chloramphenicol Rabbit Polyclonal Antibody
Supplier: Biorbyt
ORB1849-100 ORB1849-200
BIRBORB1849-100EA 0 EUR
BIRBORB1849-100 BIRBORB1849-200
Anti-Chloramphenicol Rabbit Polyclonal Antibody
Antibodies
ELISA: 1:500 (based on 0.5 mg/ml)
Type: Primary
Antigen: Chloramphenicol
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now