Anti-SMARCA2 Rabbit Polyclonal Antibody
Supplier: Biorbyt
ORB2171-100 ORB2171-200
BIRBORB2171-100EA 0 EUR
BIRBORB2171-100 BIRBORB2171-200
Anti-SMARCA2 Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:100-500; Immunochemistry: 1:50-250 (based on 0.5 mg/ml)
Type: Primary
Antigen: SMARCA2 (SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily a, member 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human