Anti-SLU7 Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB2572-100 ORB2572-200
BIRBORB2572-100EA 0 EUR
BIRBORB2572-100 BIRBORB2572-200
Anti-SLU7 Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:100-500; Immunochemistry: 1:50-250 (based on 0.5 mg/ml)

Type: Primary
Antigen: SLU7 (SLU7 splicing factor homolog (S. cerevisiae))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR