Anti-RCHY1 Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB2868-100 ORB2868-200
BIRBORB2868-100EA 0 EUR
BIRBORB2868-100 BIRBORB2868-200
Anti-RCHY1 Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:100-500; Immunochemistry: 1:50-250 (based on 0.5 mg/ml)

Type: Primary
Antigen: RCHY1 (ring finger and CHY zinc finger domain containing 1, E3 ubiquitin protein ligase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR