Anti-BAG1 Rabbit polyclonal antibody

Supplier: Biorbyt
ORB4642-100 ORB4642-200
BIRBORB4642-100EA 0 EUR
BIRBORB4642-100 BIRBORB4642-200
Anti-BAG1 Rabbit polyclonal antibody
Antibodies
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)

Type: Primary
Antigen: BAG1 (BCL2-associated athanogene)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR