Anti-TKT Rabbit Polyclonal Antibody
Supplier: Biorbyt
ORB4644-100 ORB4644-200
BIRBORB4644-100EA 0 EUR
BIRBORB4644-100 BIRBORB4644-200
Anti-TKT Rabbit Polyclonal Antibody
Antibodies
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)
Type: Primary
Antigen: TKT (transketolase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Plant