Anti-ACVR1B Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB5724-100 ORB5724-200
BIRBORB5724-100EA 0 EUR
BIRBORB5724-100 BIRBORB5724-200
Anti-ACVR1B Rabbit Polyclonal Antibody
Antibodies
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)

Type: Primary
Antigen: ACVR1B (activin A receptor, type IB)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR