Anti-C Reactive Protein Mouse Monoclonal Antibody

Supplier: Biorbyt
ORB10457-100 ORB10457-200
BIRBORB10457-100EA 0 EUR
BIRBORB10457-100 BIRBORB10457-200
Anti-C Reactive Protein Mouse Monoclonal Antibody
Antibodies
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)

Type: Primary
Antigen: C Reactive Protein
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR