Anti-DUT Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB10556-100 ORB10556-200
BIRBORB10556-100EA 0 EUR
BIRBORB10556-100 BIRBORB10556-200
Anti-DUT Rabbit Polyclonal Antibody
Antibodies
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)

Type: Primary
Antigen: DUT (deoxyuridine triphosphatase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR